Assay Details
| Gene Symbol: tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102830.1 | NP_001096300.1 | 8 - 9 | 1274 | 73 | |
| XM_012967881.2 | XP_012823335.1 | 7 - 8 | 1229 | 73 | ||
| GenBank mRNA | BC135630.1 | - | 8 - 9 | 1274 | 73 | |
| CT025340.2 | - | 6 - 7 | 1329 | 73 | ||
More Information
Set Membership: |
|

tmlhe
Genomic Map