Assay Details
| Gene Symbol: loxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102831.1 | NP_001096301.1 | 2 - 3 | 1550 | 64 | |
| GenBank mRNA | BC135631.1 | - | 2 - 3 | 1550 | 64 | |
More Information
Set Membership: |
|

loxl1
Genomic Map