Assay Details
| Gene Symbol: gnpda2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102854.1 | NP_001096324.1 | 5 - 6 | 675 | 66 | |
| GenBank mRNA | BC135724.1 | - | 5 - 6 | 675 | 66 | |
More Information
Set Membership: |
|

gnpda2
Genomic Map