Assay Details
| Gene Symbol: oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102858.1 | NP_001096328.1 | 5 - 6 | 521 | 71 | |
| GenBank mRNA | BC135746.1 | - | 5 - 6 | 521 | 71 | |
More Information
Set Membership: |
|

oxnad1
Genomic Map