Assay Details
| Gene Symbol: fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102863.1 | NP_001096333.1 | 2 - 3 | 1034 | 65 | |
| GenBank mRNA | BC135763.1 | - | 2 - 3 | 1034 | 65 | |
More Information
Set Membership: |
|

fam155b
Genomic Map