Assay Details
| Gene Symbol: ppp1r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102872.1 | NP_001096342.1 | 2 - 3 | 327 | 70 | |
| GenBank mRNA | BC135788.1 | - | 2 - 3 | 327 | 70 | |
More Information
Set Membership: |
|

ppp1r1b
Genomic Map