Assay Details
| Gene Symbol: rap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102887.1 | NP_001096357.1 | 8 - 9 | 898 | 83 | |
| XM_012964180.2 | XP_012819634.1 | 7 - 8 | 922 | 83 | ||
| GenBank mRNA | BC135848.2 | - | 8 - 9 | 898 | 83 | |
| CT025271.2 | - | 6 - 7 | 622 | 83 | ||
More Information
Set Membership: |
|

rap1a
Genomic Map