Assay Details
| Gene Symbol: fam19a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102899.1 | NP_001096369.1 | 3 - 4 | 706 | 66 | |
| GenBank mRNA | BC135900.1 | - | 3 - 4 | 706 | 66 | |
More Information
Set Membership: |
|

fam19a5
Genomic Map