Assay Details
| Gene Symbol: gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102987.1 | NP_001096457.1 | 2 - 3 | 359 | 65 | |
| XM_012967890.2 | XP_012823344.1 | 3 - 4 | 455 | 65 | ||
| GenBank mRNA | BC136180.1 | - | 2 - 3 | 359 | 65 | |
| CT030368.1 | - | 343 | 65 | |||
More Information
Set Membership: |
|

gskip
Genomic Map