Assay Details
| Gene Symbol: tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103016.1 | NP_001096486.1 | 4 - 5 | 208 | 83 | |
| GenBank mRNA | BC135327.1 | - | 4 - 5 | 208 | 83 | |
More Information
Set Membership: |
|

tnni1
Genomic Map