Assay Details
| Gene Symbol: fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103030.1 | NP_001096500.1 | 3 - 4 | 452 | 79 | |
| GenBank mRNA | BC135249.1 | - | 3 - 4 | 452 | 79 | |
More Information
Set Membership: |
|

fbxo28
Genomic Map