Assay Details
| Gene Symbol: chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103033.1 | NP_001096503.1 | 1 - 2 | 48 | 62 | |
| XM_018093766.1 | XP_017949255.1 | 1 - 2 | 113 | 62 | ||
| GenBank mRNA | BC135347.1 | - | 1 - 2 | 48 | 62 | |
More Information
Set Membership: |
|

chrng
Genomic Map