Assay Details
| Gene Symbol: mad2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103058.1 | NP_001096528.1 | 2 - 3 | 255 | 81 | |
| GenBank mRNA | BC135782.1 | - | 2 - 3 | 255 | 81 | |
| CT025283.2 | - | 2 - 3 | 211 | 81 | ||
More Information
Set Membership: |
|

mad2l1
Genomic Map