Assay Details
| Gene Symbol: nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103064.1 | NP_001096534.1 | 3 - 4 | 342 | 78 | |
| XM_012969994.2 | XP_012825448.1 | 3 - 4 | 418 | 78 | ||
| XM_018097028.1 | XP_017952517.1 | 3 - 4 | 720 | 78 | ||
| GenBank mRNA | BC135883.1 | - | 3 - 4 | 342 | 78 | |
More Information
Set Membership: |
|

nubp2
Genomic Map