Assay Details
| Gene Symbol: selenot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203537.3 | NP_988868.2 | 1 - 2 | 216 | 57 | |
| GenBank mRNA | BC059764.1 | - | 1 - 2 | 149 | 57 | |
| CF217382.1 | - | 1 - 2 | 244 | 57 | ||
| CR760355.2 | - | 1 - 2 | 161 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059764.1, CR760355.2 transcripts.
Set Membership: |
|

selenot
Genomic Map