Assay Details
| Gene Symbol: hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203811.3 | NP_989142.1 | 2 - 3 | 117 | 67 | |
| XM_018089418.1 | XP_017944907.1 | 3 - 4 | 212 | 67 | ||
| GenBank mRNA | BC061310.1 | - | 2 - 3 | 117 | 67 | |
More Information
Set Membership: |
|

hadhb
Genomic Map