Assay Details
| Gene Symbol: nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103077.1 | NP_001096547.1 | 4 - 5 | 645 | 115 | |
| XM_012967366.2 | XP_012822820.1 | 3 - 4 | 581 | 115 | ||
| XM_018094656.1 | XP_017950145.1 | 4 - 5 | 693 | 115 | ||
| XM_018094672.1 | XP_017950161.1 | 3 - 4 | 585 | 115 | ||
| XM_018094682.1 | XP_017950171.1 | 3 - 4 | 579 | 115 | ||
| XM_018094695.1 | XP_017950184.1 | 3 - 4 | 582 | 115 | ||
| XM_018094708.1 | XP_017950197.1 | 3 - 4 | 580 | 115 | ||
| XM_018094742.1 | XP_017950231.1 | 3 - 4 | 577 | 115 | ||
| GenBank mRNA | BC136181.1 | - | 4 - 5 | 645 | 115 | |
More Information
Set Membership: |
|

nmrk1
Genomic Map