Assay Details
| Gene Symbol: ca14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110051.1 | NP_001103521.1 | 7 - 8 | 873 | 77 | |
| GenBank mRNA | BC154079.1 | - | 7 - 8 | 873 | 77 | |
More Information
Set Membership: |
|

ca14
Genomic Map