Assay Details
| Gene Symbol: synj2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112913.1 | NP_001106384.1 | 2 - 3 | 259 | 63 | |
| GenBank mRNA | BC155513.1 | - | 2 - 3 | 259 | 63 | |
More Information
Set Membership: |
|

synj2bp
Genomic Map