Assay Details
| Gene Symbol: rwdd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112978.1 | NP_001106449.1 | 1 - 2 | 231 | 101 | |
| XM_012962656.2 | XP_012818110.1 | 2 - 3 | 336 | 101 | ||
| GenBank mRNA | BC153741.1 | - | 1 - 2 | 231 | 101 | |
More Information
Set Membership: |
|

rwdd2a
Genomic Map