Assay Details
| Gene Symbol: noxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112986.1 | NP_001106457.1 | 4 - 5 | 490 | 95 | |
| GenBank mRNA | BC154067.1 | - | 4 - 5 | 490 | 95 | |
More Information
Set Membership: |
|

noxo1
Genomic Map