Assay Details
| Gene Symbol: dlgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112987.1 | NP_001106458.1 | 6 - 7 | 1667 | 65 | |
| XM_012956345.2 | XP_012811799.1 | 5 - 6 | 1793 | 65 | ||
| XM_012956347.2 | XP_012811801.1 | 4 - 5 | 1708 | 65 | ||
| XM_018091059.1 | XP_017946548.1 | 5 - 6 | 1745 | 65 | ||
| XM_018091060.1 | XP_017946549.1 | 5 - 6 | 1793 | 65 | ||
| GenBank mRNA | BC154068.1 | - | 6 - 7 | 1667 | 65 | |
More Information
Set Membership: |
|

dlgap3
Genomic Map