Assay Details
| Gene Symbol: anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113009.1 | NP_001106480.1 | 12 - 13 | 1567 | 61 | |
| GenBank mRNA | BC154104.1 | - | 12 - 13 | 1567 | 61 | |
| CU464097.1 | - | 1 - 2 | 147 | 61 | ||
More Information
Set Membership: |
|

anks1a
Genomic Map