Assay Details
| Gene Symbol: tspan2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113061.1 | NP_001106532.1 | 3 - 4 | 300 | 65 | |
| GenBank mRNA | BC154902.1 | - | 3 - 4 | 300 | 65 | |
More Information
Set Membership: |
|

tspan2
Genomic Map