Assay Details
| Gene Symbol: alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113083.1 | NP_001106554.1 | 4 - 5 | 629 | 65 | |
| XM_012962660.2 | XP_012818114.1 | 4 - 5 | 501 | 65 | ||
| GenBank mRNA | BC154933.1 | - | 4 - 5 | 629 | 65 | |
More Information
Set Membership: |
|

alg3
Genomic Map