Assay Details
| Gene Symbol: trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113087.1 | NP_001106558.1 | 3 - 4 | 737 | 86 | |
| XM_018090348.1 | XP_017945837.1 | 4 - 5 | 1425 | 86 | ||
| XM_018090349.1 | XP_017945838.1 | 3 - 4 | 1581 | 86 | ||
| GenBank mRNA | BC154941.1 | - | 3 - 4 | 737 | 86 | |
More Information
Set Membership: |
|

trim25
Genomic Map