Assay Details
| Gene Symbol: pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113114.1 | NP_001106585.1 | 7 - 8 | 1744 | 85 | |
| XM_012958506.2 | XP_012813960.1 | 7 - 8 | 1801 | 85 | ||
| XM_012958507.2 | XP_012813961.1 | 6 - 7 | 2205 | 85 | ||
| XM_012958508.2 | XP_012813962.1 | 7 - 8 | 1787 | 85 | ||
| XM_012958509.1 | XP_012813963.1 | 7 - 8 | 1795 | 85 | ||
| GenBank mRNA | BC136054.1 | - | 6 - 7 | 1999 | 85 | |
| BC155407.1 | - | 7 - 8 | 1744 | 85 | ||
More Information
Set Membership: |
|

pus7l
Genomic Map