Assay Details
| Gene Symbol: afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113152.2 | NP_001106623.1 | 5 - 6 | 416 | 62 | |
| XM_012971514.2 | XP_012826968.1 | 6 - 7 | 497 | 62 | ||
| XM_012971515.2 | XP_012826969.1 | 6 - 7 | 497 | 62 | ||
| GenBank mRNA | BC155482.1 | - | 5 - 6 | 417 | 62 | |
More Information
Set Membership: |
|

afmid
Genomic Map