Assay Details
| Gene Symbol: mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113154.1 | NP_001106625.1 | 2 - 3 | 179 | 68 | |
| GenBank mRNA | BC155491.1 | - | 2 - 3 | 179 | 68 | |
More Information
Set Membership: |
|

mat1a
Genomic Map