Assay Details
| Gene Symbol: nr1h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113687.1 | NP_001107159.1 | 4 - 5 | 759 | 64 | |
| XM_012952859.1 | XP_012808313.1 | 5 - 6 | 1054 | 64 | ||
| XM_012952882.1 | XP_012808336.1 | 5 - 6 | 1061 | 64 | ||
| GenBank mRNA | BC157257.1 | - | 4 - 5 | 759 | 64 | |
| BC170929.1 | - | 4 - 5 | 759 | 64 | ||
| BC170931.1 | - | 4 - 5 | 759 | 64 | ||
More Information
Set Membership: |
|

nr1h5
Genomic Map