Assay Details
| Gene Symbol: eef1akmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113847.1 | NP_001107319.1 | 3 - 4 | 320 | 85 | |
| XM_012956383.2 | XP_012811837.1 | 3 - 4 | 375 | 85 | ||
| XM_012956384.2 | XP_012811838.1 | 3 - 4 | 337 | 85 | ||
| XM_012956386.2 | XP_012811840.1 | 3 - 4 | 390 | 85 | ||
| XM_012956387.1 | XP_012811841.1 | 3 - 4 | 271 | 85 | ||
| GenBank mRNA | BC155986.1 | - | 3 - 4 | 320 | 85 | |
| BC156004.1 | - | 3 - 4 | 320 | 85 | ||
| BC171256.1 | - | 3 - 4 | 309 | 85 | ||
More Information
Set Membership: |
|

eef1akmt1
Genomic Map