Assay Details
| Gene Symbol: ugt1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113894.1 | NP_001107366.1 | 1 - 2 | 1099 | 72 | |
| XM_012970020.2 | XP_012825474.1 | 2 - 3 | 1216 | 72 | ||
| XM_012970022.2 | XP_012825476.1 | 1 - 2 | 1217 | 72 | ||
| GenBank mRNA | BC157226.1 | - | 1 - 2 | 1099 | 72 | |
| BC171210.1 | - | 1 - 2 | 970 | 72 | ||
| BC171213.1 | - | 1 - 2 | 970 | 72 | ||
More Information
Set Membership: |
|

ugt1a6
Genomic Map