Assay Details
| Gene Symbol: ugt1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113894.1 | NP_001107366.1 | 2 - 3 | 1228 | 67 | |
| XM_012970020.2 | XP_012825474.1 | 3 - 4 | 1345 | 67 | ||
| XM_012970022.2 | XP_012825476.1 | 2 - 3 | 1346 | 67 | ||
| GenBank mRNA | BC157226.1 | - | 2 - 3 | 1228 | 67 | |
| BC171210.1 | - | 2 - 3 | 1099 | 67 | ||
| BC171213.1 | - | 2 - 3 | 1099 | 67 | ||
More Information
Set Membership: |
|

ugt1a6
Genomic Map