Assay Details
| Gene Symbol: ugt1a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113894.1 | NP_001107366.1 | 4 - 5 | 1543 | 67 | |
| XM_012970020.2 | XP_012825474.1 | 5 - 6 | 1660 | 67 | ||
| GenBank mRNA | BC157226.1 | - | 4 - 5 | 1543 | 67 | |
| BC171210.1 | - | 4 - 5 | 1414 | 67 | ||
| BC171213.1 | - | 4 - 5 | 1414 | 67 | ||
More Information
Set Membership: |
|

ugt1a6
Genomic Map