Assay Details
| Gene Symbol: ada.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113897.1 | NP_001107369.1 | 4 - 5 | 477 | 94 | |
| XM_012971519.2 | XP_012826973.1 | 4 - 5 | 499 | 94 | ||
| GenBank mRNA | BC157229.1 | - | 4 - 5 | 477 | 94 | |
| BC171134.1 | - | 3 - 4 | 336 | 94 | ||
| BC171136.1 | - | 3 - 4 | 336 | 94 | ||
More Information
Set Membership: |
|

ada.2
Genomic Map