Assay Details
| Gene Symbol: ada.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113897.1 | NP_001107369.1 | 7 - 8 | 788 | 58 | |
| XM_012971519.2 | XP_012826973.1 | 7 - 8 | 810 | 58 | ||
| GenBank mRNA | BC157229.1 | - | 7 - 8 | 788 | 58 | |
| BC171134.1 | - | 6 - 7 | 647 | 58 | ||
| BC171136.1 | - | 6 - 7 | 647 | 58 | ||
More Information
Set Membership: |
|

ada.2
Genomic Map