Assay Details
| Gene Symbol: dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113898.1 | NP_001107370.1 | 1 - 2 | 944 | 96 | |
| GenBank mRNA | BC157230.1 | - | 1 - 2 | 944 | 96 | |
| BC171240.1 | - | 1 - 2 | 933 | 96 | ||
| BC171242.1 | - | 1 - 2 | 933 | 96 | ||
More Information
Set Membership: |
|

dnajc22
Genomic Map