Assay Details
| Gene Symbol: smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114202.1 | NP_001107674.1 | 1 - 2 | 437 | 71 | |
| GenBank mRNA | BC086613.1 | - | 1 - 2 | 151 | 71 | |
| BC154887.1 | - | 1 - 2 | 182 | 71 | ||
| CR848311.2 | - | 1 - 2 | 437 | 71 | ||
More Information
Set Membership: |
|

smdt1
Genomic Map