Assay Details
| Gene Symbol: smdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114202.1 | NP_001107674.1 | 2 - 3 | 567 | 80 | |
| GenBank mRNA | BC086613.1 | - | 2 - 3 | 281 | 80 | |
| BC154887.1 | - | 2 - 3 | 312 | 80 | ||
| CR848311.2 | - | 2 - 3 | 567 | 80 | ||
More Information
Set Membership: |
|

smdt1
Genomic Map