Assay Details
| Gene Symbol: il17d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114247.1 | NP_001107719.1 | 2 - 3 | 344 | 92 | |
| XM_012956404.2 | XP_012811858.1 | 2 - 3 | 3148 | 92 | ||
| GenBank mRNA | BC158482.1 | - | 2 - 3 | 344 | 92 | |
More Information
Set Membership: |
|

il17d
Genomic Map