Assay Details
| Gene Symbol: znf652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114255.1 | NP_001107727.1 | 2 - 3 | 839 | 69 | |
| GenBank mRNA | BC158164.1 | - | 2 - 3 | 839 | 69 | |
More Information
Set Membership: |
|

znf652
Genomic Map