Assay Details
| Gene Symbol: setmar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114263.1 | NP_001107735.1 | 1 - 2 | 140 | 60 | |
| GenBank mRNA | BC158450.1 | - | 1 - 2 | 140 | 60 | |
More Information
Set Membership: |
|

setmar
Genomic Map