Assay Details
| Gene Symbol: fgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114490.1 | NP_001107962.1 | 6 - 7 | 841 | 71 | |
| GenBank mRNA | BC094532.1 | - | 6 - 7 | 821 | 71 | |
| BC158991.1 | - | 6 - 7 | 841 | 71 | ||
More Information
Set Membership: |
|

fgb
Genomic Map