Assay Details
| Gene Symbol: prpf39.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122801.1 | NP_001116273.1 | 12 - 13 | 2320 | 71 | |
| XM_012967892.2 | XP_012823346.1 | 14 - 15 | 2065 | 71 | ||
| XM_012967893.2 | XP_012823347.1 | 13 - 14 | 2091 | 71 | ||
| GenBank mRNA | BC124575.1 | - | 12 - 13 | 2320 | 71 | |
| BC135138.1 | - | 6 - 7 | 789 | 71 | ||
More Information
Set Membership: |
|

prpf39.1
Genomic Map