Assay Details
| Gene Symbol: prpf39.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122801.1 | NP_001116273.1 | 9 - 10 | 1942 | 100 | |
| XM_012967892.2 | XP_012823346.1 | 11 - 12 | 1687 | 100 | ||
| XM_012967893.2 | XP_012823347.1 | 10 - 11 | 1713 | 100 | ||
| GenBank mRNA | BC124575.1 | - | 9 - 10 | 1942 | 100 | |
| BC135138.1 | - | 3 - 4 | 411 | 100 | ||
More Information
Set Membership: |
|

prpf39.1
Genomic Map