Assay Details
| Gene Symbol: nlk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123413.1 | NP_001116885.1 | 10 - 11 | 1692 | 69 | |
| GenBank mRNA | BC159140.1 | - | 10 - 11 | 1692 | 69 | |
More Information
Set Membership: |
|

nlk
Genomic Map