Assay Details
| Gene Symbol: zranb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123428.1 | NP_001116900.1 | 2 - 3 | 964 | 74 | |
| XM_018095246.1 | XP_017950735.1 | 3 - 4 | 1119 | 74 | ||
| XM_018095247.1 | XP_017950736.1 | 2 - 3 | 856 | 74 | ||
| XM_018095248.1 | XP_017950737.1 | 2 - 3 | 907 | 74 | ||
| XM_018095249.1 | XP_017950738.1 | 2 - 3 | 1083 | 74 | ||
| GenBank mRNA | BC161086.1 | - | 2 - 3 | 964 | 74 | |
More Information
Set Membership: |
|

zranb1
Genomic Map