Assay Details
| Gene Symbol: klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123440.1 | NP_001116912.1 | 4 - 5 | 1126 | 61 | |
| XM_012967977.2 | XP_012823431.1 | 4 - 5 | 1453 | 61 | ||
| XM_012967978.2 | XP_012823432.1 | 4 - 5 | 1467 | 61 | ||
| XM_018096145.1 | XP_017951634.1 | 5 - 6 | 1542 | 61 | ||
| XM_018096146.1 | XP_017951635.1 | 3 - 4 | 1344 | 61 | ||
| GenBank mRNA | BC161356.1 | - | 4 - 5 | 1126 | 61 | |
More Information
Set Membership: |
|

klf8
Genomic Map