Assay Details
| Gene Symbol: zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123454.1 | NP_001116926.1 | 5 - 6 | 638 | 60 | |
| XM_012966029.2 | XP_012821483.1 | 5 - 6 | 673 | 60 | ||
| XM_012966030.2 | XP_012821484.1 | 5 - 6 | 667 | 60 | ||
| XM_018095250.1 | XP_017950739.1 | 5 - 6 | 637 | 60 | ||
| XM_018095251.1 | XP_017950740.1 | 5 - 6 | 636 | 60 | ||
| XM_018095252.1 | XP_017950741.1 | 5 - 6 | 637 | 60 | ||
| XM_018095253.1 | XP_017950742.1 | 5 - 6 | 637 | 60 | ||
| XM_018095254.1 | XP_017950743.1 | 5 - 6 | 636 | 60 | ||
| XM_018095255.1 | XP_017950744.1 | 5 - 6 | 635 | 60 | ||
| GenBank mRNA | BC159030.1 | - | 5 - 6 | 638 | 60 | |
More Information
Set Membership: |
|

zfyve27
Genomic Map