Assay Details
| Gene Symbol: il5ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123462.1 | NP_001116934.1 | 11 - 12 | 1404 | 81 | |
| XM_012960586.2 | XP_012816040.1 | 11 - 12 | 1404 | 81 | ||
| XM_012960587.2 | XP_012816041.1 | 12 - 13 | 1526 | 81 | ||
| XM_012960588.1 | XP_012816042.1 | 11 - 12 | 1374 | 81 | ||
| XM_012960589.2 | XP_012816043.1 | 11 - 12 | 1373 | 81 | ||
| GenBank mRNA | BC161054.1 | - | 11 - 12 | 1404 | 81 | |
More Information
Set Membership: |
|

il5ra
Genomic Map